4-Chloro-2-(hydroxymethyl)benzimidazole

Lieferant: Apollo Scientific

OR471359-1G
APOSOR471359-1GEA 804 EUR
APOSOR471359-1G
4-Chloro-2-(hydroxymethyl)benzimidazole
4-Chloro-2-(hydroxymethyl)benzimidazole
Formel: C₈H₇ClN₂O
Molecular Weight: 182,607 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20763992
CAS-Nummer: 1263060-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR