2-(Hydroxymethyl)-4,6-dimethylbenzimidazole

Lieferant: Apollo Scientific

OR471361-1G
APOSOR471361-1GEA 1010 EUR
APOSOR471361-1G
2-(Hydroxymethyl)-4,6-dimethylbenzimidazole
2-(Hydroxymethyl)-4,6-dimethylbenzimidazole
Formel: C₁₀H₁₂N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD23724090
CAS-Nummer: 1049110-09-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR