5,6-Dichloro-2-(hydroxymethyl)benzimidazole

Lieferant: Apollo Scientific

OR471362-1G
APOSOR471362-1GEA 713 EUR
APOSOR471362-1G
5,6-Dichloro-2-(hydroxymethyl)benzimidazole
5,6-Dichloro-2-(hydroxymethyl)benzimidazole
Formel: C₈H₆Cl₂N₂O
Molecular Weight: 217,052 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09880338
CAS-Nummer: 6478-80-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR