(7-Methyl-1H-benzimidazol-2-yl)methanol

Lieferant: Apollo Scientific

OR471364-5G
APOSOR471364-5GEA 383 EUR
APOSOR471364-5G
(7-Methyl-1H-benzimidazol-2-yl)methanol
(7-Methyl-1H-benzimidazol-2-yl)methanol
Formel: C₉H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09971880
CAS-Nummer: 191794-20-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR