5-Hydroxy-2-(hydroxymethyl)benzimidazole

Lieferant: Apollo Scientific

OR471365-1G
APOSOR471365-1GEA 601 EUR
APOSOR471365-1G
5-Hydroxy-2-(hydroxymethyl)benzimidazole
5-Hydroxy-2-(hydroxymethyl)benzimidazole
Formel: C₈H₈N₂O₂
Molecular Weight: 164,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03170139
CAS-Nummer: 886493-60-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR