2-(3-Chloropropyl)-6-methoxyquinazoline

Lieferant: Apollo Scientific

OR471366-1G
APOSOR471366-1GEA 855 EUR
APOSOR471366-1G
2-(3-Chloropropyl)-6-methoxyquinazoline
2-(3-Chloropropyl)-6-methoxyquinazoline
Formel: C₁₂H₁₃ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD28977361
CAS-Nummer: 2006276-98-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR