2-(Chloromethyl)-6-methoxyquinazoline

Lieferant: Apollo Scientific

OR471368-1G
APOSOR471368-1GEA 855 EUR
APOSOR471368-1G
2-(Chloromethyl)-6-methoxyquinazoline
2-(Chloromethyl)-6-methoxyquinazoline
Formel: C₁₀H₉ClN₂O
Molecular Weight: 208,644 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977359
CAS-Nummer: 1936095-76-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR