6-Bromo-2-(3-chloropropyl)quinazoline

Lieferant: Apollo Scientific

OR471369-1G
APOSOR471369-1GEA 1010 EUR
APOSOR471369-1G
6-Bromo-2-(3-chloropropyl)quinazoline
6-Bromo-2-(3-chloropropyl)quinazoline
Formel: C₁₁H₁₀BrClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28977358
CAS-Nummer: 2006277-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR