6-Bromo-2-(2-chloroethyl)quinazoline

Lieferant: Apollo Scientific

OR471370-1G
APOSOR471370-1GEA 1010 EUR
APOSOR471370-1G
6-Bromo-2-(2-chloroethyl)quinazoline
6-Bromo-2-(2-chloroethyl)quinazoline
Formel: C₁₀H₈BrClN₂
Molecular Weight: 271,541 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977357
CAS-Nummer: 1934521-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR