6-Bromo-2-(chloromethyl)quinazoline

Lieferant: Apollo Scientific

OR471371-1G
APOSOR471371-1GEA 1010 EUR
APOSOR471371-1G
6-Bromo-2-(chloromethyl)quinazoline
6-Bromo-2-(chloromethyl)quinazoline
Formel: C₉H₆BrClN₂
Molecular Weight: 257,514 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977356
CAS-Nummer: 1936538-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR