7-Bromo-2-(3-chloropropyl)quinazoline

Lieferant: Apollo Scientific

OR471372-1G
APOSOR471372-1GEA 1010 EUR
APOSOR471372-1G
7-Bromo-2-(3-chloropropyl)quinazoline
7-Bromo-2-(3-chloropropyl)quinazoline
Formel: C₁₁H₁₀BrClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28977352
CAS-Nummer: 2006277-09-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR