7-Bromo-2-(2-chloroethyl)quinazoline

Lieferant: Apollo Scientific

OR471373-1G
APOSOR471373-1GEA 1010 EUR
APOSOR471373-1G
7-Bromo-2-(2-chloroethyl)quinazoline
7-Bromo-2-(2-chloroethyl)quinazoline
Formel: C₁₀H₈BrClN₂
Molecular Weight: 271.541 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977351
CAS-Nummer: 1935427-67-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR