7-Bromo-2-(chloromethyl)quinazoline

Lieferant: Apollo Scientific

OR471374-1G
APOSOR471374-1GEA 1610 EUR
APOSOR471374-1G
7-Bromo-2-(chloromethyl)quinazoline
7-Bromo-2-(chloromethyl)quinazoline
Formel: C₉H₆BrClN₂
Molecular Weight: 257,514 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28977350
CAS-Nummer: 1936495-91-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR