2-(3-Chloropropyl)-6,7-dimethoxyquinazoline
Lieferant: Apollo Scientific
OR471375-1G
APOSOR471375-1GEA
1010
EUR
APOSOR471375-1G
2-(3-Chloropropyl)-6,7-dimethoxyquinazoline
2-(3-Chloropropyl)-6,7-dimethoxyquinazoline
Formel:
C₁₃H₁₅ClN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28977349 CAS-Nummer: 2006277-49-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...