2-(2-Chloroethyl)-6,7-dimethoxyquinazoline

Lieferant: Apollo Scientific

OR471376-1G
APOSOR471376-1GEA 1010 EUR
APOSOR471376-1G
2-(2-Chloroethyl)-6,7-dimethoxyquinazoline
2-(2-Chloroethyl)-6,7-dimethoxyquinazoline
Formel: C₁₂H₁₃ClN₂O₂
Molecular Weight: 252,697 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977348
CAS-Nummer: 2006277-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR