2-(Chloromethyl)-6,7-dimethoxyquinazoline

Lieferant: Apollo Scientific

OR471377-1G
APOSOR471377-1GEA 1010 EUR
APOSOR471377-1G
2-(Chloromethyl)-6,7-dimethoxyquinazoline
2-(Chloromethyl)-6,7-dimethoxyquinazoline
Formel: C₁₁H₁₁ClN₂O₂
Molecular Weight: 238,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977347
CAS-Nummer: 2006276-97-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR