6-Chloro-2-(2-chloroethyl)quinazoline
Lieferant: Apollo Scientific
OR471379-1G
APOSOR471379-1GEA
1010
EUR
APOSOR471379-1G
6-Chloro-2-(2-chloroethyl)quinazoline
6-Chloro-2-(2-chloroethyl)quinazoline
Formel:
C₁₀H₈Cl₂N₂ Molecular Weight: 227,09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28977345 CAS-Nummer: 1936054-22-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...