2-(3-Chloropropyl)quinazoline

Lieferant: Apollo Scientific

OR471381-1G
APOSOR471381-1GEA 1010 EUR
APOSOR471381-1G
2-(3-Chloropropyl)quinazoline
2-(3-Chloropropyl)quinazoline
Formel: C₁₁H₁₁ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28977343
CAS-Nummer: 1934527-68-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR