2-(2-Chloroethyl)quinazoline

Lieferant: Apollo Scientific

OR471382-1G
APOSOR471382-1GEA 1010 EUR
APOSOR471382-1G
2-(2-Chloroethyl)quinazoline
2-(2-Chloroethyl)quinazoline
Formel: C₁₀H₉ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28977342
CAS-Nummer: 1934503-72-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR