Tetrakis(4-nitrophenyl)methane

Lieferant: Apollo Scientific
Achtung

OR471383-5G OR471383-1G
APOSOR471383-5GEA 1220 EUR
APOSOR471383-5G APOSOR471383-1G
Tetrakis(4-nitrophenyl)methane
Tetrakis(4-nitrophenyl)methane
Formel: C₂₅H₁₆N₄O₈
Lagertemperatur: Raumtemperatur
MDL: MFCD12546931
CAS-Nummer: 60532-62-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR