Ethyl 2,4,6-Trichloronicotinate

Lieferant: Apollo Scientific

Synonyme: Ethyl 2,4,6-trichloronicotinate

OR471384-1G
APOSOR471384-1GEA 1010 EUR
APOSOR471384-1G
Ethyl 2,4,6-Trichloronicotinate
Ethyl 2,4,6-Trichloronicotinate
Formel: C₈H₆Cl₃NO₂
Molecular Weight: 254,498 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28503376
CAS-Nummer: 1934657-78-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR