4,7-Dichloro-3-nitroquinoline
Lieferant: Apollo Scientific
OR471385-1G
APOSOR471385-1GEA
1830
EUR
APOSOR471385-1G
4,7-Dichloro-3-nitroquinoline
4,7-Dichloro-3-nitroquinoline
Formel:
C₉H₄Cl₂N₂O₂ Molecular Weight: 243,046 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17170308 CAS-Nummer: 22931-74-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...