4-Chloro-5-nitroquinazoline

Lieferant: Apollo Scientific

OR471386-1G
APOSOR471386-1GEA 1330 EUR
APOSOR471386-1G
4-Chloro-5-nitroquinazoline
4-Chloro-5-nitroquinazoline
Formel: C₈H₄ClN₃O₂
Molecular Weight: 209,589 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18448299
CAS-Nummer: 75224-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR