Methyl-4-chlorchinolin-2-carboxylat
Lieferant: Apollo Scientific
OR471387-1G
OR471387-250MG
APOSOR471387-1GEA
810
EUR
APOSOR471387-1G
APOSOR471387-250MG
Methyl-4-chlorchinolin-2-carboxylat
Methyl-4-chlorchinolin-2-carboxylat
Formel:
C₁₁H₈ClNO₂ Molecular Weight: 221.639 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14706446 CAS-Nummer: 114935-92-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...