Methyl-4-chlorchinolin-2-carboxylat

Lieferant: Apollo Scientific

OR471387-1G OR471387-250MG
APOSOR471387-1GEA 810 EUR
APOSOR471387-1G APOSOR471387-250MG
Methyl-4-chlorchinolin-2-carboxylat
Methyl-4-chlorchinolin-2-carboxylat
Formel: C₁₁H₈ClNO₂
Molecular Weight: 221.639 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14706446
CAS-Nummer: 114935-92-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR