Methyl 3-[Hydroxy(3-pyridyl)methyl]isoxazole-4-carboxylate

Lieferant: Apollo Scientific

OR471395-1G
APOSOR471395-1GEA 1220 EUR
APOSOR471395-1G
Methyl 3-[Hydroxy(3-pyridyl)methyl]isoxazole-4-carboxylate
Methyl 3-[Hydroxy(3-pyridyl)methyl]isoxazole-4-carboxylate
Formel: C₁₁H₁₀N₂O₄
Molecular Weight: 234,208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977403
CAS-Nummer: 2006276-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR