3-[(3-Pyridyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid

Lieferant: Apollo Scientific

OR471396-1G
APOSOR471396-1GEA 1220 EUR
APOSOR471396-1G
3-[(3-Pyridyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
3-[(3-Pyridyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
Formel: C₁₅H₁₆N₂O₅
Lagertemperatur: Raumtemperatur
MDL: MFCD28977402
CAS-Nummer: 2006277-53-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR