3-[Hydroxy(3-pyridyl)methyl]isoxazole-4-carboxylic acid

Lieferant: Apollo Scientific

OR471397-1G
APOSOR471397-1GEA 1220 EUR
APOSOR471397-1G
3-[Hydroxy(3-pyridyl)methyl]isoxazole-4-carboxylic acid
3-[Hydroxy(3-pyridyl)methyl]isoxazole-4-carboxylic acid
Formel: C₁₀H₈N₂O₄
Molecular Weight: 220,182 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977401
CAS-Nummer: 2006277-29-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR