Ethyl 3-[Hydroxy(4-nitrophenyl)methyl]isoxazole-4-carboxylate

Lieferant: Apollo Scientific

OR471398-1G
APOSOR471398-1GEA 1220 EUR
APOSOR471398-1G
Ethyl 3-[Hydroxy(4-nitrophenyl)methyl]isoxazole-4-carboxylate
Ethyl 3-[Hydroxy(4-nitrophenyl)methyl]isoxazole-4-carboxylate
Formel: C₁₃H₁₂N₂O₆
Molecular Weight: 292,244 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977396
CAS-Nummer: 2006277-72-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR