Methyl 3-[Hydroxy(4-nitrophenyl)methyl]isoxazole-4-carboxylate
Lieferant: Apollo Scientific
OR471399-1G
APOSOR471399-1GEA
1220
EUR
APOSOR471399-1G
Methyl 3-[Hydroxy(4-nitrophenyl)methyl]isoxazole-4-carboxylate
Methyl 3-[Hydroxy(4-nitrophenyl)methyl]isoxazole-4-carboxylate
Formel:
C₁₂H₁₀N₂O₆ Molecular Weight: 278,218 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28977395 CAS-Nummer: 2006277-45-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...