3-[(4-Nitrophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid

Lieferant: Apollo Scientific

OR471400-1G
APOSOR471400-1GEA 1220 EUR
APOSOR471400-1G
3-[(4-Nitrophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
3-[(4-Nitrophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
Formel: C₁₆H₁₆N₂O₇
Molecular Weight: 348,307 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977394
CAS-Nummer: 2006277-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR