3-[Hydroxy(4-nitrophenyl)methyl]isoxazole-4-carboxylic acid
Lieferant: Apollo Scientific
OR471401-1G
APOSOR471401-1GEA
1220
EUR
APOSOR471401-1G
3-[Hydroxy(4-nitrophenyl)methyl]isoxazole-4-carboxylic acid
3-[Hydroxy(4-nitrophenyl)methyl]isoxazole-4-carboxylic acid
Formel:
C₁₁H₈N₂O₆ Molecular Weight: 264,191 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28977393 CAS-Nummer: 2006278-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...