Ethyl 3-[(4-Bromophenyl)(hydroxy)methyl]isoxazole-4-carboxylate
Lieferant: Apollo Scientific
OR471402-1G
APOSOR471402-1GEA
1220
EUR
APOSOR471402-1G
Ethyl 3-[(4-Bromophenyl)(hydroxy)methyl]isoxazole-4-carboxylate
Ethyl 3-[(4-Bromophenyl)(hydroxy)methyl]isoxazole-4-carboxylate
Formel:
C₁₃H₁₂BrNO₄ Molecular Weight: 326,143 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28977388 CAS-Nummer: 2006277-63-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...