Methyl 3-[(4-Bromophenyl)(hydroxy)methyl]isoxazole-4-carboxylate

Lieferant: Apollo Scientific

OR471403-1G
APOSOR471403-1GEA 1220 EUR
APOSOR471403-1G
Methyl 3-[(4-Bromophenyl)(hydroxy)methyl]isoxazole-4-carboxylate
Methyl 3-[(4-Bromophenyl)(hydroxy)methyl]isoxazole-4-carboxylate
Formel: C₁₂H₁₀BrNO₄
Molecular Weight: 312,116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977387
CAS-Nummer: 2006277-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR