3-[(4-Bromophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid

Lieferant: Apollo Scientific

OR471404-1G
APOSOR471404-1GEA 1220 EUR
APOSOR471404-1G
3-[(4-Bromophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
3-[(4-Bromophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
Formel: C₁₆H₁₆BrNO₅
Molecular Weight: 382,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977386
CAS-Nummer: 2006277-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR