3-[(4-Bromophenyl)(hydroxy)methyl]isoxazole-4-carboxylic acid

Lieferant: Apollo Scientific

OR471405-1G
APOSOR471405-1GEA 1220 EUR
APOSOR471405-1G
3-[(4-Bromophenyl)(hydroxy)methyl]isoxazole-4-carboxylic acid
3-[(4-Bromophenyl)(hydroxy)methyl]isoxazole-4-carboxylic acid
Formel: C₁₁H₈BrNO₄
Molecular Weight: 298,09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977385
CAS-Nummer: 2006276-91-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR