Ethyl 3-[(4-Chlorophenyl)(hydroxy)methyl]isoxazole-4-carboxylate

Lieferant: Apollo Scientific

OR471406-1G
APOSOR471406-1GEA 1220 EUR
APOSOR471406-1G
Ethyl 3-[(4-Chlorophenyl)(hydroxy)methyl]isoxazole-4-carboxylate
Ethyl 3-[(4-Chlorophenyl)(hydroxy)methyl]isoxazole-4-carboxylate
Formel: C₁₃H₁₂ClNO₄
Molecular Weight: 281,692 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977384
CAS-Nummer: 2006277-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR