Methyl 3-[(4-Chlorophenyl)(hydroxy)methyl]isoxazole-4-carboxylate

Lieferant: Apollo Scientific

OR471407-1G
APOSOR471407-1GEA 1220 EUR
APOSOR471407-1G
Methyl 3-[(4-Chlorophenyl)(hydroxy)methyl]isoxazole-4-carboxylate
Methyl 3-[(4-Chlorophenyl)(hydroxy)methyl]isoxazole-4-carboxylate
Formel: C₁₂H₁₀ClNO₄
Molecular Weight: 267,665 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977383
CAS-Nummer: 2006277-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR