3-[(4-Chlorophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
Lieferant: Apollo Scientific
OR471408-1G
APOSOR471408-1GEA
1220
EUR
APOSOR471408-1G
3-[(4-Chlorophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
3-[(4-Chlorophenyl)[(2-tetrahydropyranyl)oxy]methyl]isoxazole-4-carboxylic acid
Formel:
C₁₆H₁₆ClNO₅ Molecular Weight: 337,755 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28977382 CAS-Nummer: 2006277-13-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...