3-[(4-Chlorophenyl)(hydroxy)methyl]isoxazole-4-carboxylic acid

Lieferant: Apollo Scientific

OR471409-1G
APOSOR471409-1GEA 1220 EUR
APOSOR471409-1G
3-[(4-Chlorophenyl)(hydroxy)methyl]isoxazole-4-carboxylic acid
3-[(4-Chlorophenyl)(hydroxy)methyl]isoxazole-4-carboxylic acid
Formel: C₁₁H₈ClNO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD28977381
CAS-Nummer: 2006277-75-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR