Ethyl 3-[Hydroxy(phenyl)methyl]isoxazole-4-carboxylate

Lieferant: Apollo Scientific

OR471410-1G
APOSOR471410-1GEA 1220 EUR
APOSOR471410-1G
Ethyl 3-[Hydroxy(phenyl)methyl]isoxazole-4-carboxylate
Ethyl 3-[Hydroxy(phenyl)methyl]isoxazole-4-carboxylate
Formel: C₁₃H₁₃NO₄
Molecular Weight: 247,247 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977380
CAS-Nummer: 2006277-85-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR