Methyl 3-[Hydroxy(phenyl)methyl]isoxazole-4-carboxylate

Lieferant: Apollo Scientific

OR471411-1G
APOSOR471411-1GEA 1220 EUR
APOSOR471411-1G
Methyl 3-[Hydroxy(phenyl)methyl]isoxazole-4-carboxylate
Methyl 3-[Hydroxy(phenyl)methyl]isoxazole-4-carboxylate
Formel: C₁₂H₁₁NO₄
Molecular Weight: 233,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977379
CAS-Nummer: 2006277-12-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR