3-[Hydroxy(phenyl)methyl]isoxazole-4-carboxylic acid

Lieferant: Apollo Scientific

OR471413-1G
APOSOR471413-1GEA 1220 EUR
APOSOR471413-1G
3-[Hydroxy(phenyl)methyl]isoxazole-4-carboxylic acid
3-[Hydroxy(phenyl)methyl]isoxazole-4-carboxylic acid
Formel: C₁₁H₉NO₄
Molecular Weight: 219,193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977377
CAS-Nummer: 2006277-34-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR