Methyl 3-Ethylisoxazole-4-carboxylate

Lieferant: Apollo Scientific

OR471414-5G
APOSOR471414-5GEA 804 EUR
APOSOR471414-5G
Methyl 3-Ethylisoxazole-4-carboxylate
Methyl 3-Ethylisoxazole-4-carboxylate
Formel: C₇H₉NO₃
Molecular Weight: 155.151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977376
CAS-Nummer: 1402812-89-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR