3-Ethylisoxazole-4-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR471416-1G
APOSOR471416-1GEA
713
EUR
APOSOR471416-1G
3-Ethylisoxazole-4-carboxylic acid
3-Ethylisoxazole-4-carboxylic acid
Formel:
C₆H₇NO₃ Molecular Weight: 141,125 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19229970 CAS-Nummer: 639523-12-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...