(R)-3-Phenyl-2-(1-pyrrolidinyl)propanoic acid

Lieferant: Apollo Scientific

OR471419-1G
APOSOR471419-1GEA 804 EUR
APOSOR471419-1G
(R)-3-Phenyl-2-(1-pyrrolidinyl)propanoic acid
(R)-3-Phenyl-2-(1-pyrrolidinyl)propanoic acid
Formel: C₁₃H₁₇NO₂
Molecular Weight: 219,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977414
CAS-Nummer: 2006286-95-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR