(R)-4-Methyl-2-(1-pyrrolidinyl)pentanoic acid

Lieferant: Apollo Scientific

OR471420-1G
APOSOR471420-1GEA 804 EUR
APOSOR471420-1G
(R)-4-Methyl-2-(1-pyrrolidinyl)pentanoic acid
(R)-4-Methyl-2-(1-pyrrolidinyl)pentanoic acid
Formel: C₁₀H₁₉NO₂
Molecular Weight: 185,263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28977413
CAS-Nummer: 2006286-93-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR