(S)-4-Methyl-2-(1-pyrrolidinyl)pentanoic acid
Lieferant: Apollo Scientific
OR471421-1G
APOSOR471421-1GEA
804
EUR
APOSOR471421-1G
(S)-4-Methyl-2-(1-pyrrolidinyl)pentanoic acid
(S)-4-Methyl-2-(1-pyrrolidinyl)pentanoic acid
Formel:
C₁₀H₁₉NO₂ Molecular Weight: 185,263 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20656288 CAS-Nummer: 753435-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...