(R)-3-Methyl-2-(1-pyrrolidinyl)butyric acid
Lieferant: Apollo Scientific
OR471422-1G
APOSOR471422-1GEA
804
EUR
APOSOR471422-1G
(R)-3-Methyl-2-(1-pyrrolidinyl)butyric acid
(R)-3-Methyl-2-(1-pyrrolidinyl)butyric acid
Formel:
C₉H₁₇NO₂ Molecular Weight: 171,237 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28977412 CAS-Nummer: 1932443-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...