(S)-3-Methyl-2-(1-pyrrolidinyl)butyric acid

Lieferant: Apollo Scientific

OR471423-1G
APOSOR471423-1GEA 804 EUR
APOSOR471423-1G
(S)-3-Methyl-2-(1-pyrrolidinyl)butyric acid
(S)-3-Methyl-2-(1-pyrrolidinyl)butyric acid
Formel: C₉H₁₇NO₂
Molecular Weight: 171,237 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20656283
CAS-Nummer: 777839-82-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR